missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEK 293T cells Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27555-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is designed to detect the proteins of the HEK 293T cells.
HEK 293T cell line, an important variant of HEK 293 cell line, which contains the SV40 Large T-antigen that allows for episomal replication of transfected plasmids containing the SV40 origin of replication. HEK 293T cells are usually used for plasmids transfection and temporal expression of desired gene products.Specifications
| HEK 293T cells | |
| Polyclonal | |
| Unconjugated | |
| 293T, HEK 293T cells, HEK293T | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 1.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Rabbit | |
| 150 μL | |
| Primary | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction