missing translation for 'onlineSavingsMsg'
Learn More

Hecogenin (>80%), TRC

Product Code. 30392827
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392827 250 mg
30406025 50 mg
30395537 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392827 Supplier Toronto Research Chemicals Supplier No. TRCH129945250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 467-55-0
Molecular Formula C27 H42 O4
Molecular Weight (g/mol) 430.62
Synonym Spirostan-12-one, 3-hydroxy-, (3β,5α,25R)-, 5α,25D-Spirostan-12-one, 3β-hydroxy- (7CI), 5α-Spirostan-12-one, 3β-hydroxy-, (25R)- (8CI), (+)-Hecogenin, (25R)-3β-Hydroxy-5α-spirostan-12-one, (3β,5α,25R)-3-Hydroxyspirostan-12-one, 12-Oxotigogenin, 25(R)-3β-Hydroxyspirostan-12-one, 3β-Hydroxy-5α-hecogenin, Gekogenin, Hecogenin, Hekogenin, Hocogenin, NSC 115921
SMILES C[C@@H]1CC[C@@]2(OC1)O[C@H]3C[C@H]4[C@@H]5CC[C@H]6C[C@@H](O)CC[C@]6(C)[C@H]5CC(=O)[C@]4(C)[C@H]3[C@@H]2C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C27H42O4/c1-15-7-10-27(30-14-15)16(2)24-22(31-27)12-21-19-6-5-17-11-18(28)8-9-25(17,3)20(19)13-23(29)26(21,24)4/h15-22,24,28H,5-14H2,1-4H3/t15-,16+,17+,18+,19-,20+,21+,22+,24+,25+,26-,27-/m1/s1
Formula Weight 430.3083
Chemical Name or Material Hecogenin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.