CAS RN 467-55-0
IUPAC Name:
(1'R,2R,2'S,4'S,5R,7'S,8'R,9'S,12'S,13'S,16'S,18'S)-16'-hydroxy-5,7',9',13'-tetramethyl-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan]-10'-one
Synonyms:
hecogenin hocogenin
Molecular Weight (g/mol):
430.63
Molecular Formula:
C27H42O4
InChi Key:
QOLRLLFJMZLYQJ-LOBDNJQFSA-N
SMILES:
C[C@H]1[C@H]2[C@H](C[C@H]3[C@@H]4CC[C@H]5C[C@@H](O)CC[C@]5(C)[C@H]4CC(=O)[C@]23C)O[C@]11CC[C@@H](C)CO1
1
–
1
of
1
results
1
Hecogenin (>80%), TRC
CAS: 467-55-0 Molecular Formula: C27 H42 O4 Molecular Weight (g/mol): 430.62 Synonym: Spirostan-12-one, 3-hydroxy-, (3β,5α,25R)-,5α,25D-Spirostan-12-one, 3β-hydroxy- (7CI),5α-Spirostan-12-one, 3β-hydroxy-, (25R)- (8CI),(+)-Hecogenin,(25R)-3β-Hydroxy-5α-spirostan-12-one,(3β,5α,25R)-3-Hydroxyspirostan-12-one,12-Oxotigogenin,25(R)-3β-Hydroxyspirostan-12-one,3β-Hydroxy-5α-hecogenin,Gekogenin,Hecogenin,Hekogenin,Hocogenin,NSC 115921 SMILES: C[C@@H]1CC[C@@]2(OC1)O[C@H]3C[C@H]4[C@@H]5CC[C@H]6C[C@@H](O)CC[C@]6(C)[C@H]5CC(=O)[C@]4(C)[C@H]3[C@@H]2C