missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Heating/Cooling Mini Shakers

Product Code. p-6731002 Shop All Fisherbrand Products
Change view
Click to view available options
Electrical Requirements:
230 V,1 A, 450 W
Plug Type:
EU, UK, CN Plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Plug Type unitSize
15544080 230 V,1 A, 450 W EU, UK, CN Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15544080
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for analyses that requires a stable, controlled temperature.

  • Microprocessor control -The variable speed microprocessor control provides consistent, uniform shaking action. Microprocessor will display last set-point and will restart if power is interrupted.
  • PID temperature controller - Maintains precise temperature control from 10°C below ambient to 65°C. Easy-to-use controls allow users to adjust temperature in 1°C increments.
  • Triple eccentric drive - Permanently lubricated ball bearings and maintenance free, brushless DC motor provide reliable service and continuous duty operation.
  • LED display- Touch pad controls with easy-to-read, independent LED displays for temperature, speed and time allow operator to view all settings at once. Provides repeatable and accurate results every time and is easily visible across lab benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off.
  • Temperature calibration mode - Allows user to calibrate unit to an external temperature device.
  • Applications - Cell and bacterial cultures, hybridizations, and enzyme reactions
TRUSTED_SUSTAINABILITY

Specifications

Orbit 3 mm
Speed Control Accuracy ±2% of set speed up to 999 rpm ±5% 1000 to 1200 rpm
Speed Range 150 to 1200 rpm (Microplate), 100 to 600 rpm (Modular Block)
Electrical Requirements 230 V,1 A, 450 W
Plug Type EU, UK, CN Plug
Dimensions (LxW) Platform 7 x 5.5 in. (180 x 140 mm)
Dimensions (L x W x H) 16.3 x 11.5 x 12 in. (415 x 293 x 308 mm)
Alarms Audible and Visual
Display LED
Humidity Range Maximum 80%
Timer Range 1 sec. to 9999 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.