missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Grab Rack™

Product Code. 11701426 Shop All Fisherbrand Products
Change view
Click to view available options
Positions:
50 Wells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions unitSize
11701426 50 Wells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

Not for cleaning in autoclave

Product Code. 11701426

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Not for cleaning in autoclave

Dump the contents, not your tubes. Fisherbrand™ Grab Rack™ uses silicon grips to securely hold fifty 5 to 10mL tubes in place, so you can empty all of the tubes at once for greater efficiency. Insert or remove upright tubes into the dual-level rack using gentle pressure.

  • Rack features silicon well grips that keep tubes securely in place
  • Double-layer ABS rack design holds tubes upright
  • Tubes do not fall out when rack is turned over, so you can discard all the contents at once
  • Tubes insert and extract with a gentle push or pull
  • Features a 5 x 10 well array; no assembly required

Specifications

Includes Silicon Grip
Color White
Material ABS Plastic
Positions 50 Wells
For Use With (Equipment) 5 to 10mL tubes
Size 259 x 127 x 49 mm
Dimensions (L x W x H) 259 x 127 x 49mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.