Learn More
Grant Instruments™ Heat Exchange Cooling Coil
Can be attached to a cold water supply or refrigerated circulator. Grant Instruments™ Heat Exchange Coil maintains bath down to 2°C above the temperature of the coolant.
Brand: Grant Instruments™ CW26
Code : NEW
Additional Details : Weight : 2.00000kg
Description
Can be attached to a cold water supply or refrigerated circulator. Grant Instruments™ Heat Exchange Coil maintains bath down to 2°C above the temperature of the coolant.
- Stainless steel heat exchange coil that can be attached to cold water supply or a refrigerated circulator
- Can be used to maintain a water bath temperature that is 2°C above the temperature of the coolant used
- Compatible with OLS 26 Aqua Pro
Specifications
OLS26 | |
Heat Exchange Coil |