Learn More
Grant Instruments™ Heat Exchange Cooling Coil
Can be attached to a cold water supply or refrigerated circulator. Grant Instruments™ Heat Exchange Coil maintains bath down to 2°C above the temperature of the coolant.
£238.50 - £471.70
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15345872
|
Grant Instruments™
CW26 |
OLS26 |
£471.70
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11486487
|
Grant Instruments™
CW5 |
T100, TC120, TX150, TXF200 |
£238.50
Each |
In Stock
Log in to see stock. |
|||||
Description
Can be attached to a cold water supply or refrigerated circulator. Grant Instruments™ Heat Exchange Coil maintains bath down to 2°C above the temperature of the coolant.
- Stainless steel heat exchange coil that can be attached to cold water supply or a refrigerated circulator
- Can be used to maintain a water bath temperature that is 2°C above the temperature of the coolant used
- Compatible with OLS 26 Aqua Pro
Specifications
Heat Exchange Coil |