missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ HDPE Measuring Scoop

Product Code. 10097141
Change view
Click to view available options
Capacity (Metric):
15 mL
65 mL
110 mL
350 mL
Unit Size:
Each
Pack of 12
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10097141 110 mL Pack of 12
10502322 350 mL Pack of 6
10047520 350 mL Each
10390621 65 mL Pack of 12
10106550 15 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10097141 Supplier VITLAB™ Supplier No. 40393
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these corrosion resistant scoops for applications in the food industry. VITLAB™ HDPE Measuring Scoops are lighter than metal, don’t dent and feature good chemical resistance.

  • Conical in shape with tapered filling edge

Specifications

Capacity (Metric) 110 mL
Color Natural
Product Type Measuring Scoop
For Use With (Application) Suitable as weighing
Material HDPE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.