missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ HDPE Measuring Scoop
Description
- Conical in shape with tapered filling edge
Specifications
Specifications
| Capacity (Metric) | 350 mL |
| Color | Natural |
| Product Type | Measuring Scoop |
| For Use With (Application) | Suitable as weighing |
| Material | HDPE |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?