missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDLBP Recombinant Rabbit Monoclonal Antibody (23GB3975), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | HDLBP |
|---|---|
| Clone | 23GB3975 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089862
![]() |
Invitrogen™
MA552650 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089112
|
Invitrogen™
MA552651 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDLBP Recombinant Monoclonal antibody specifically detects HDLBP in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| HDLBP | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q00341, Q8VDJ3, Q9Z1A6 | |
| 110611, 3069, 64474 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3975 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110005P14Rik; AA960365; AI118566; D1Ertd101e; HBP; HDL binding protein; HDL-binding protein; HDLBP; Hemopexin; high density lipoprotein (HDL) binding protein; high density lipoprotein binding protein; high density lipoprotein binding protein (vigilin); high density lipoprotein-binding protein; Hpx; Hpxn; hx; lipoprotein-binding protein; PRO2900; VGL; vigilin | |
| A synthesized peptide derived from human HDLBP (1200-1268AA). | |
| Primary | |
| -20°C | |
| HDLBP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title