missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDLBP Recombinant Rabbit Monoclonal Antibody (23GB3975), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552650
This item is not returnable.
View return policy
Description
High density lipoprotein-binding protein, also known as vigilin, is a 110-kD protein that specifically binds HDL molecules and may function in the removal of excess cellular cholesterol.
Specifications
| HDLBP | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HDLBP | |
| 1110005P14Rik; AA960365; AI118566; D1Ertd101e; HBP; HDL binding protein; HDL-binding protein; HDLBP; Hemopexin; high density lipoprotein (HDL) binding protein; high density lipoprotein binding protein; high density lipoprotein binding protein (vigilin); high density lipoprotein-binding protein; Hpx; Hpxn; hx; lipoprotein-binding protein; PRO2900; VGL; vigilin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 110611, 3069, 64474 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3975 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q00341, Q8VDJ3, Q9Z1A6 | |
| HDLBP | |
| A synthesized peptide derived from human HDLBP (1200-1268AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction