missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDJ2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£164.00 - £375.00
Specifications
| Antigen | HDJ2 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18341056
|
Novus Biologicals
NBP3-03520-20UL |
20 μL |
£164.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18393417
|
Novus Biologicals
NBP3-03520-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDJ2 Polyclonal specifically detects HDJ2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| HDJ2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| dj-2, DjA1, DnaJ (Hsp40) homolog, subfamily A, member 1, dnaJ homolog subfamily A member 1, DnaJ protein homolog 2, DNAJ2, hDJ-2, HDJ2, Heat shock 40 kDa protein 4, Heat shock protein J2, heat shock protein, DNAJ-like 2, HSDJ, HSJ2, HSJ-2, HSPF4hdj-2, Human DnaJ protein 2, NEDD7, neural precursor cell expressed, developmentally down-regulated 7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 178-397 of human HDJ2 (NP_001530.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
| PBS with 50% glycerol, pH7.3. | |
| 3301 | |
| IgG | |
| Affinity purified |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto