missing translation for 'onlineSavingsMsg'
Learn More

HDJ2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18341056 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341056 20 μL 20µL
18393417 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18341056 Supplier Novus Biologicals Supplier No. NBP30352020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HDJ2 Polyclonal specifically detects HDJ2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDJ2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias dj-2, DjA1, DnaJ (Hsp40) homolog, subfamily A, member 1, dnaJ homolog subfamily A member 1, DnaJ protein homolog 2, DNAJ2, hDJ-2, HDJ2, Heat shock 40 kDa protein 4, Heat shock protein J2, heat shock protein, DNAJ-like 2, HSDJ, HSJ2, HSJ-2, HSPF4hdj-2, Human DnaJ protein 2, NEDD7, neural precursor cell expressed, developmentally down-regulated 7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 178-397 of human HDJ2 (NP_001530.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3301
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.