missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDJ2 Antibody (23C6), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | HDJ2 |
|---|---|
| Clone | 23C6 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043479
|
Novus Biologicals
NBP3-26616-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043253
|
Novus Biologicals
NBP3-26616-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDJ2 Monoclonal antibody specifically detects HDJ2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
| HDJ2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| dj-2, DjA1, DnaJ (Hsp40) homolog, subfamily A, member 1, dnaJ homolog subfamily A member 1, DnaJ protein homolog 2, DNAJ2, hDJ-2, HDJ2, Heat shock 40 kDa protein 4, Heat shock protein J2, heat shock protein, DNAJ-like 2, HSDJ, HSJ2, HSJ-2, HSPF4hdj-2, Human DnaJ protein 2, NEDD7, neural precursor cell expressed, developmentally down-regulated 7 | |
| A synthesized peptide derived from Human HDJ2 [UniProt P31689] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 23C6 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3301 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title