missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDJ2 Antibody (23C6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26616-100ul
This item is not returnable.
View return policy
Description
HDJ2 Monoclonal antibody specifically detects HDJ2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Specifications
| HDJ2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| 23C6 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| dj-2, DjA1, DnaJ (Hsp40) homolog, subfamily A, member 1, dnaJ homolog subfamily A member 1, DnaJ protein homolog 2, DNAJ2, hDJ-2, HDJ2, Heat shock 40 kDa protein 4, Heat shock protein J2, heat shock protein, DNAJ-like 2, HSDJ, HSJ2, HSJ-2, HSPF4hdj-2, Human DnaJ protein 2, NEDD7, neural precursor cell expressed, developmentally down-regulated 7 | |
| A synthesized peptide derived from Human HDJ2 [UniProt P31689] | |
| 100 μL | |
| DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
| 3301 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction