missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17548-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is a rabbit polyclonal antibody raised against full length HDAC8 of human origin.
In the intact cell, DNA closely associates with histones and other nuclear proteins to form chromatin. The remodeling of chromatin is believed to be a critical component of transcriptional regulation and a major source of this remodeling is brought about by the acetylation of nucleosomal histones. Acetylation of lysine residues in the amino terminal tail domain of histone results in an allosteric change in the nucleosomal conformation and an increased accessibility to transcription factors by DNA. Conversely, the deacetylation of histones is associated with transcriptional silencing. Several mammalian proteins have been identified as nuclear histone acetylases, including GCN5, PCAF (p300/CBP-associated factor), p300/CBP, HAT1 and the TFIID subunit TAF II p250. Mammalian HDAC8, isolated from human kidney, is a histone deacetylase that shares homology to other HDACs but has different tissue distribution. HDAC8 is localized to the nucleus and plays a role in the development of a broad range of tissues and in the etiology of cancer.Specifications
HDAC8 | |
Polyclonal | |
Unconjugated | |
HDAC8 | |
HD8, HDAC8, HDACL1, histone deacetylase 8, RPD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
363481, 55869, 70315 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B1WC68, Q8VH37, Q9BY41 | |
Hdac8 | |
HDAC8 Fusion Protein Ag11696 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |