missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | HDAC8 |
---|---|
Concentration | 0.14 mg/mL |
Applications | Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877454
|
Proteintech
17548-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867454
|
Proteintech
17548-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is a rabbit polyclonal antibody raised against full length HDAC8 of human origin.
In the intact cell, DNA closely associates with histones and other nuclear proteins to form chromatin. The remodeling of chromatin is believed to be a critical component of transcriptional regulation and a major source of this remodeling is brought about by the acetylation of nucleosomal histones. Acetylation of lysine residues in the amino terminal tail domain of histone results in an allosteric change in the nucleosomal conformation and an increased accessibility to transcription factors by DNA. Conversely, the deacetylation of histones is associated with transcriptional silencing. Several mammalian proteins have been identified as nuclear histone acetylases, including GCN5, PCAF (p300/CBP-associated factor), p300/CBP, HAT1 and the TFIID subunit TAF II p250. Mammalian HDAC8, isolated from human kidney, is a histone deacetylase that shares homology to other HDACs but has different tissue distribution. HDAC8 is localized to the nucleus and plays a role in the development of a broad range of tissues and in the etiology of cancer.Specifications
HDAC8 | |
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
B1WC68, Q8VH37, Q9BY41 | |
363481, 55869, 70315 | |
HDAC8 Fusion Protein Ag11696 | |
Primary | |
-20°C | |
HDAC8 |
0.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HD8, HDAC8, HDACL1, histone deacetylase 8, RPD3 | |
Hdac8 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |