missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7 Rabbit anti-Human, Mouse, Rat, Clone: 6J2N9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | HDAC7 |
|---|---|
| Clone | 6J2N9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18371684
|
Novus Biologicals
NBP3-15457-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18362765
|
Novus Biologicals
NBP3-15457-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC7 Monoclonal antibody specifically detects HDAC7 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| HDAC7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 51564 | |
| IgG | |
| Affinity purified |
| 6J2N9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 24-198 of human HDAC7 (Q8WUI4). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title