missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7, p Ser318 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£160.00 - £329.00
Specifications
| Antigen | HDAC7 (p Ser318) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.1 - 1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18012947
|
Novus Biologicals
NBP2-03978SS |
0.025 mg |
£160.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18044904
|
Novus Biologicals
NBP2-03978 |
0.05 mg |
£329.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC7 Polyclonal specifically detects HDAC7 in Human, Bovine, Canine, Equine, Primate, Zebrafish samples. It is validated for Western Blot.Specifications
| HDAC7 (p Ser318) | |
| Western Blot 0.1 - 1 ug/mL | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases, Phospho Specific | |
| PBS with 0.02% Sodium Azide | |
| DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 | |
| HDAC7 | |
| IgG | |
| Affinity Purified | |
| Reacts with Chimpanzee, Cow, Dog, Horse, Human, New World Monkey and Zebrafish. Predicted to react with Mouse, Rat and Chicken. |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Bovine, Canine, Equine, Primate, Zebrafish | |
| NP_055522 | |
| 51564 | |
| A synthetic peptide containing phospho-serine 318 of human HDAC7 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title