missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7, p Ser318 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-03978SS
This item is not returnable.
View return policy
Description
HDAC7 Polyclonal specifically detects HDAC7 in Human, Bovine, Canine, Equine, Primate, Zebrafish samples. It is validated for Western Blot.
Specifications
| HDAC7 (p Ser318) | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reacts with Chimpanzee, Cow, Dog, Horse, Human, New World Monkey and Zebrafish. Predicted to react with Mouse, Rat and Chicken. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 0.1 - 1 ug/mL | |
| NP_055522 | |
| HDAC7 | |
| A synthetic peptide containing phospho-serine 318 of human HDAC7 was used as the immunogen. | |
| 0.025 mg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases, Phospho Specific | |
| 51564 | |
| Human, Bovine, Canine, Equine, Primate, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction