missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-45611
This item is not returnable.
View return policy
Description
HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HDAC6 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol with 0.01% Thimerosal | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| Rabbit | |
| 131 kDa | |
| 0.1 mg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases | |
| 10013 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 20 ug/mL | |
| Q9Z2V5 | |
| HDAC6 | |
| Synthetic peptide mapping to the N-terminus of mouse HDAC-6. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody does not cross-react with other HDAC proteins including HDAC1, 2, 3, 4, 5, 7, and 8. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction