missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £448.00
Specifications
| Antigen | HDAC6 |
|---|---|
| Dilution | Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 20 ug/mL |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18206905
|
Novus Biologicals
NBP1-45611-0.025MG |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18237162
|
Novus Biologicals
NBP1-45611 |
0.1 mg |
£448.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| HDAC6 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Rat, Mouse | |
| Q9Z2V5 | |
| 10013 | |
| Synthetic peptide mapping to the N-terminus of mouse HDAC-6. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 131 kDa |
| Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 20 ug/mL | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases | |
| PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol with 0.01% Thimerosal | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| HDAC6 | |
| IgG | |
| Affinity Purified | |
| The antibody does not cross-react with other HDAC proteins including HDAC1, 2, 3, 4, 5, 7, and 8. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title