missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Antibody (2H12), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | HDAC6 |
|---|---|
| Clone | 2H12 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30029634
|
Novus Biologicals
NBP3-26236-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30031902
|
Novus Biologicals
NBP3-26236-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC6 Monoclonal antibody specifically detects HDAC6 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunoprecipitationSpecifications
| HDAC6 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| A synthesized peptide derived from Human HDAC6 [UniProt Q9UBN7] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2H12 | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 10013 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title