missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Antibody (2H12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26236-50ul
This item is not returnable.
View return policy
Description
HDAC6 Monoclonal antibody specifically detects HDAC6 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Specifications
| HDAC6 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| 2H12 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 | |
| EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
| A synthesized peptide derived from Human HDAC6 [UniProt Q9UBN7] | |
| 50 μL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Neuroscience | |
| 10013 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction