missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ HCS NuclearMask™ Stains

Product Code. 10726885
Click to view available options
Color:
Blue
Far-Red, Deep Red
Red
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10726885

Brand: Molecular Probes™ H10325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Easily identify cell nuclei and other features of interest during high-content screening (HCS) assays with the versatile HCS NuclearMask Blue, Red, and Deep Red stains, which can be used with live and fixed cells and in multiparametric analyses.

Easily identify cell nuclei in high-content screening (HCS) assays with the versatile HCS NuclearMask Blue, Red, and Deep Red stains, which survive standard formaldehyde-based fixation and detergent-based permeabilization methods and can be applied to live and fixed cells. Because their dyes are easily discriminated from other fluorophores, the HCS NuclearMask stains can be used in multiparametric analyses. They can also label the entire cell (i.e., cytoplasm and nucleus) and provide an accurate backdrop against which to assess the features of interest.

The versatile HCS NuclearMask Blue, HCS NuclearMask Red, and HCS NuclearMask Deep Red stains can be used to measure DNA content and perform cell demarcation with formaldehyde-fixed cells on high content imaging and analysis (HCS) platforms. HCS NuclearMask Blue, HCS NuclearMask Red, and HCS NuclarMask Deep Red stains have excitation/emission maxima of 355/460 nm, 622/645 nm, and 638/686 nm, respectively, when bound to DNA. HCS NuclearMask Blue stain is easily discriminated from visible- and near-IR wavelength fluorophores, HCS NuclearMask Red stain is easily discriminated from green- and even orange-fluorescent fluorophores, and HCS NuclearMask Deep Red is easily discriminated from visible wavelength fluorophores when used in multiparametric analyses.

If total cell staining is desired, HCS CellMask stains label the entire cell (cytoplasm and nucleus) and provide an accurate backdrop against which the features of interest can be assessed. HCS CellMask stains are available in a choice of colors for multiplexing flexibility. Sufficient volume quantities are provided to stain ten 96-well plates using an assay volume of 100 μL per well.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Content And Storage Includes 65 μL of 2000X concentrate, providing sufficient material to stain a total of ten 96-well microplates.

Store at 2–6°C and protect from light.

Excitation Wavelength Range 350/451
For Use With (Equipment) High Content Instrument
Quantity 65 μL
Detection Method Fluorescence
Description HCS NuclearMask™ Blue Stain
Emission Visible
Product Line NuclearMask
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Stain
SubCellular Localization Nucleic Acids, Nucleus
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.