Learn More
Invitrogen™ HCS LIVE/DEAD™ Green Kit
Description
The HCS LIVE/DEAD™ Green Kit measures cytotoxicity by high content analysis with the Image-iT™ DEAD Green™ viability stain—a non-fluorescent and cell impermeant compound that exhibits a strong fluorescent enhancement upon binding to DNA. Drugs and test compounds leading to serious cell injuries, including plasma membrane permeability, allow entry of the Image-iT™ DEAD Green™ viability stain. This two-color fluorescence assay also employs the use of a cell permeant nuclear segmentation tool that stains both live and dead cells, reflecting the total cell number within the sample. For total cell staining, the kit includes both a blue-fluorescent Hoechst 33342 and a near infrared-fluorescent, HCS NuclearMask™ Deep Red reagent. Either of these segmentation tools may be used with Image-iT™ DEAD Green™ viability stain for excellent discrimination of live and dead cells that survives fixation and permeabilization for combination with additional parameters of cytotoxicity or other biomarkers. The kit contains sufficient reagents for performing 2, 96-well plates.
Specifications
Specifications
| Cell Type | Eukaryotic Cells |
| Color | Green, Far-red, Blue |
| Content And Storage | Store at ≤-20°C, desiccate and protect from light. |
| Description | HCS LIVE/DEAD™ Green Kit, Blue/Far-Red/Green, 96-Well Plate Format, Fluorescent Detection Method, 350/461, 488/515, 638/686nm Excitation/Emission, Used For Cell Viability, Proliferation & Function, Cellular Imaging |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay, Cellular Imaging, High-content Screening (HCS), Cell Viability, Proliferation and Function, Cell Viability and Cytotoxicity |
| For Use With (Equipment) | Fluorescence Microscope, High Content Analysis Instrument |
| Product Type | HCS LIVE/DEAD Green Kit |
| Dye Type | HCS NuclearMask™ Deep Red stain, Image-iT™ DEAD Green viability stain, Hoechst 33342 |
| Format | 96-well plate |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.