missing translation for 'onlineSavingsMsg'
Learn More

hCG beta Mouse anti-Human, Clone: 2D6B8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60334-1-IG-150UL

 View more versions of this product

Product Code. 16877435

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Human chorionic gonadotropin (hCG) is a placental hormone that stimulates secretion of the pregcy-sustaining steroid progesterone. hCG is a member of a family of glycoprotein hormones including pituitary hormones lutropin (luteinizing hormone;LH), follitropin (follicle-stimulating hormone; FSH) and thyrotropin (thyroid-stimulating hormone; TSH). Members of this family are all composed of disulfide-rich heterodimers with two dissimilar subunits namely alpha- and beta-subunits. The alpha subunit is common to all the members of the family within a given species. The beta subunits are distinct in each of the horomones and confer the receptor and biological specificity.
Specifications

Specifications

hCG beta
Monoclonal
0.79 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P0DN86
CGB3
hCG beta Fusion Protein Ag2191
150 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
2D6B8
Unconjugated
CGB3
CG beta, CGB, CGB,hCG, CGB3, CGB5, CGB7, CGB8, hCG, hCGB
Mouse
Protein A
RUO
1082
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.