missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hCG beta Mouse anti-Human, Clone: 2D6B8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | hCG beta |
|---|---|
| Clone | 2D6B8 |
| Concentration | 0.79 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16887435
|
Proteintech
60334-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16877435
|
Proteintech
60334-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human chorionic gonadotropin (hCG) is a placental hormone that stimulates secretion of the pregcy-sustaining steroid progesterone. hCG is a member of a family of glycoprotein hormones including pituitary hormones lutropin (luteinizing hormone;LH), follitropin (follicle-stimulating hormone; FSH) and thyrotropin (thyroid-stimulating hormone; TSH). Members of this family are all composed of disulfide-rich heterodimers with two dissimilar subunits namely alpha- and beta-subunits. The alpha subunit is common to all the members of the family within a given species. The beta subunits are distinct in each of the horomones and confer the receptor and biological specificity.Specifications
| hCG beta | |
| 0.79 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CG beta, CGB, CGB,hCG, CGB3, CGB5, CGB7, CGB8, hCG, hCGB | |
| CGB3 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 2D6B8 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P0DN86 | |
| 1082 | |
| hCG beta Fusion Protein Ag2191 | |
| Primary | |
| -20°C | |
| CGB3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title