missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCAP-G Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | HCAP-G |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18375584
|
Novus Biologicals
NBP3-03775-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18316896
|
Novus Biologicals
NBP3-03775-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HCAP-G Polyclonal specifically detects HCAP-G in Human samples. It is validated for Western Blot.Specifications
| HCAP-G | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CAP-G, CAPGHCAP-G, CHCG, chromosome condensation protein G, Chromosome-associated protein G, condensin complex subunit 3, Condensin subunit CAP-G, FLJ12450, hCAP-G, Melanoma antigen NY-MEL-3, MGC126525, Non-SMC condensin I complex subunit G, non-SMC condensin I complex, subunit G, NYMEL3, NY-MEL-3, XCAP-G homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 610-700 of human HCAP-G (NP_071741.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, DNA Repair | |
| PBS with 50% glycerol, pH7.3. | |
| 64151 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title