missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCAP-G Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03775-100UL
This item is not returnable.
View return policy
Description
HCAP-G Polyclonal specifically detects HCAP-G in Human samples. It is validated for Western Blot.
Specifications
| HCAP-G | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| CAP-G, CAPGHCAP-G, CHCG, chromosome condensation protein G, Chromosome-associated protein G, condensin complex subunit 3, Condensin subunit CAP-G, FLJ12450, hCAP-G, Melanoma antigen NY-MEL-3, MGC126525, Non-SMC condensin I complex subunit G, non-SMC condensin I complex, subunit G, NYMEL3, NY-MEL-3, XCAP-G homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 610-700 of human HCAP-G (NP_071741.2). | |
| 100 μL | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, DNA Repair | |
| 64151 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction