missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAT1 Recombinant Rabbit Monoclonal Antibody (23GB2090), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | HAT1 |
|---|---|
| Clone | 23GB2090 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087570
![]() |
Invitrogen™
MA553220 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088849
|
Invitrogen™
MA553221 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HAT1 Recombinant Monoclonal antibody specifically detects HAT1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| HAT1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O14929, Q5M939, Q8BY71 | |
| 107435, 296501, 8520 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2090 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2410071B14Rik; AA536933; HAT1; Hat-1; histidine aminotransferase 1; histone acetyltransferase 1; Histone acetyltransferase type B catalytic subunit; histone aminotransferase 1; KAT1 | |
| A synthesized peptide derived from human KAT1/HAT1 (200-419AA). | |
| Primary | |
| -20°C | |
| HAT1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title