missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAT1 Recombinant Rabbit Monoclonal Antibody (23GB2090), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553221
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a type B histone acetyltransferase (HAT) that is involved in the rapid acetylation of newly synthesized cytoplasmic histones, which are in turn imported into the nucleus for de novo deposition onto nascent DNA chains. Histone acetylation, particularly of histone H4, plays an important role in replication-dependent chromatin assembly. Specifically, this HAT can acetylate soluble but not nucleosomal histone H4 at lysines 5 and 12, and to a lesser degree, histone H2A at lysine 5. Alternatively spliced transcript variants have been identified for this gene.
Specifications
| HAT1 | |
| Recombinant Monoclonal | |
| 1.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14929, Q5M939, Q8BY71 | |
| HAT1 | |
| A synthesized peptide derived from human KAT1/HAT1 (200-419AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2090 | |
| Unconjugated | |
| HAT1 | |
| 2410071B14Rik; AA536933; HAT1; Hat-1; histidine aminotransferase 1; histone acetyltransferase 1; Histone acetyltransferase type B catalytic subunit; histone aminotransferase 1; KAT1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 107435, 296501, 8520 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction