missing translation for 'onlineSavingsMsg'
Learn More

Harringtonin, TRC

Product Code. 30391099
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391099 10 mg
30402731 100 mg
30398521 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391099 Supplier Toronto Research Chemicals Supplier No. TRCH10528510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26833-85-2
Molecular Formula C28 H37 N O9
Molecular Weight (g/mol) 531.59
Synonym 3-[4-Methyl (2R)-2-hydroxy-2-(3-hydroxy-3-methylbutyl)butanedioate]cephalotaxine, 2'R-Harringtonine, NSC 124147
SMILES COC(=O)CC(O)(CCC(C)(C)O)C(=O)O[C@@H]1C2c3cc4OCOc4cc3CCN5CCC[C@@]25C=C1OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C28H37NO9/c1-26(2,32)8-9-28(33,15-22(30)35-4)25(31)38-24-21(34-3)14-27-7-5-10-29(27)11-6-17-12-19-20(37-16-36-19)13-18(17)23(24)27/h12-14,23-24,32-33H,5-11,15-16H2,1-4H3/t23?,24-,27+,28?/m0/s1
Formula Weight 531.2468
Chemical Name or Material Harringtonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.