CAS RN 26833-85-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Harringtonin, TRC
CAS: 26833-85-2 Molecular Formula: C28 H37 N O9 Molecular Weight (g/mol): 531.59 Synonym: 3-[4-Methyl (2R)-2-hydroxy-2-(3-hydroxy-3-methylbutyl)butanedioate]cephalotaxine,2'R-Harringtonine,NSC 124147 SMILES: COC(=O)CC(O)(CCC(C)(C)O)C(=O)O[C@@H]1C2c3cc4OCOc4cc3CCN5CCC[C@@]25C=C1OC
2
Harringtonine, MedChemExpress
MedChemExpress Harringtonine is a natural Cephalotaxus alkaloid that inhibits protein synthesis. Harringtonine has anti-chikungunya virus (CHIKV) activities with an EC50 of 0.24 μM.