Learn More
Hard Wood Test Tube Racks
Organize and store test tubes and samples with these stands. Hard Wood Test Tube Racks occupy only about one third of the space required by a conventional rack when dismantled for storage.
Brand:  Samco L330/22
Code : 03
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13098423
  • £93.95 / Pack of 5
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
  • Single row test tube stand
  • Hardwood top and bottom members
  • Molded polypropylene supports which clip into position
  • Packs flat for transport and storage
Specifications
Specifications
Test Tube Rack
Tubes (22 mm)
6
22 mm
6
Hardwood
5 Pack
22 mm