missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hard Wood Test Tube Racks
Organize and store test tubes and samples with these stands. Hard Wood Test Tube Racks occupy only about one third of the space required by a conventional rack when dismantled for storage.
£93.95 - £142.00
Product Code | Brand | Diameter (Metric) Cavity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Cavity | Price | Quantity & Availability | |||||
13028433
|
Samco
L330/21 |
32 mm |
£142.00
Pack of 5 |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
13008433
|
Samco
L330/25 |
10 mm |
£136.00
Pack of 5 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
13098423
|
Samco
L330/22 |
22 mm |
£93.95
Pack of 5 |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||||
Description
- Single row test tube stand
- Hardwood top and bottom members
- Molded polypropylene supports which clip into position
- Packs flat for transport and storage
Specifications
Test Tube Rack | |
Hardwood | |
5 Pack |