missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HARBI1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56801
This item is not returnable.
View return policy
Description
HARBI1 Polyclonal specifically detects HARBI1 in Human samples. It is validated for Western Blot.
Specifications
| HARBI1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C11orf77, chromosome 11 open reading frame 77, EC 3.1, FLJ32675, harbinger transposase derived 1, Harbinger transposase-derived nuclease, putative nuclease HARBI1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 283254 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96MB7 | |
| HARBI1 | |
| Synthetic peptides corresponding to C11ORF77 The peptide sequence was selected from the middle region of C11ORF77. Peptide sequence GVMGVVDCIHVAIKAPNAEDLSYVNRKGLHSLNCLMVCDIRGTLMTVETN. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Mouse: 100%; Rat: 100%; Chicken: 100%; Canine: 100%; Bovine: 100%; Human: 100%; Xenopus: 91%; blue catfish: 85%; Zebrafish: 78%;. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction