missing translation for 'onlineSavingsMsg'
Learn More

Hantaan Virus Nucleocapsid Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£150.00 - £427.00

Specifications

Antigen Hantaan Virus Nucleocapsid
Concentration 1 mg/mL
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18650337
View Documents
Novus Biologicals
NBP2-41259-0.025mg
0.025 mg
£150.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18106458
View Documents
Novus Biologicals
NBP2-41259
0.1 mg
£427.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hantaan Virus Nucleocapsid Polyclonal specifically detects Hantaan Virus Nucleocapsid in Virus samples. It is validated for ELISA.
Specifications

Specifications

Hantaan Virus Nucleocapsid
ELISA
Unconjugated
RUO
PBS with 0.02% Sodium Azide
Antibody was raised against an 18 amino acid peptide near the amino terminus of the Hantaan virus nucleocapsid. The immunogen is located within the first 50 amino acids of Hantaan Virus Nucleocapsid. Amino Acid Squence: YEKDPDELNKRTLTDREG
Primary
48 kDa
1 mg/mL
Polyclonal
Rabbit
Virus
HTNVsSgp1
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.