missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hantaan Virus Nucleocapsid Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41259-0.025mg
This item is not returnable.
View return policy
Description
Hantaan Virus Nucleocapsid Polyclonal specifically detects Hantaan Virus Nucleocapsid in Virus samples. It is validated for ELISA.
Specifications
| Hantaan Virus Nucleocapsid | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| 48 kDa | |
| 0.025 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| ELISA | |
| 1 mg/mL | |
| ELISA 1 μg/mL | |
| HTNVsSgp1 | |
| Antibody was raised against an 18 amino acid peptide near the amino terminus of the Hantaan virus nucleocapsid. The immunogen is located within the first 50 amino acids of Hantaan Virus Nucleocapsid. Amino Acid Squence: YEKDPDELNKRTLTDREG | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction