missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile
GREENER_CHOICE

Product Code. 11364295 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
20 μL
50 μL
200 μL
1000 μL
Format:
Hanging Tip Racked
Hanging Tip Racked, Cardboard
Unit Size:
Pack of 1536
Pack of 2304
Pack of 4608
Pack of 6912
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Format unitSize
11364295 200 μL Hanging Tip Racked Pack of 2304
11364285 1000 μL Hanging Tip Racked Pack of 1536
12521288 1000 μL Hanging Tip Racked, Cardboard Pack of 4608
11314295 20 μL Hanging Tip Racked Pack of 2304
11314305 50 μL Hanging Tip Racked Pack of 2304
12571318 50 μL Hanging Tip Racked, Cardboard Pack of 6912
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11364295 Supplier Axygen™ Supplier No. TT200CBKHTR
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Black tips for accurate sample delivery

  • Polypropylene
  • Volume ranging from 20 to 1000μL
  • Available with or without aerosol-resistant filters
  • Compatible with Axygen™ Tecan™/Perkin Elmer/Auroroa™ Biomed/Corbett Cas-1200 systems

Certifications

Certified DNAse-, RNAse- and Pyrogen-safe

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Black
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
For Use With (Application) For accurate sample delivery
Format Hanging Tip Racked
Tip Style Automation, Round
For Use With (Equipment) Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.