missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Clear Tips, Presterilized
GREENER_CHOICE

Product Code. 11354285 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
20 μL
50 μL
200 μL
1000 μL
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 1536
Pack of 2304
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Filtered unitSize
11354285 1000 μL Non-filtered Pack of 1536
11354295 200 μL Non-filtered Pack of 2304
11304305 50 μL Non-filtered Pack of 2304
11344305 1000 μL Filtered Pack of 1536
11384305 200 μL Filtered Pack of 2304
11364305 20 μL Filtered Pack of 2304
11304315 50 μL Filtered Pack of 2304
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11354285 Supplier Axygen™ Supplier No. TT1000CHTRS
  • ONLINE EXCLUSIVE PRICE
    £180.00
    £175.80 / Pack of 1536
    Save £4.20
    2% Off
View all promotions
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Clear tips for accurate sample delivery

Axygen™ offers a comprehensive line of tips for Tecan™, Aurora Biomed and PerkinElmer™ automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in nonsterile, sterile, non-filtered and filtered. Clear and Conductive black polypropylene tips are available in 96-well format and is also available in hanging tip rack format designed for Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head and PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head.


  • 1000 μL Clear Tips
  • 96 Tip format
  • Clear Polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Sterile
  • Compatibility: Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head
  • Certifications

    Certified DNAse-, RNAse- and Pyrogen-safe

    TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 1000 μL
    Filtered Non-filtered
    Sterility Sterile
    Color Clear
    Autoclavable Autoclavable
    Certifications/Compliance Non-ANSI
    For Use With (Application) For accurate sample delivery
    Format Rack
    Tip Style Automation, Round
    Packaging Hanging tip rack
    Quantity 16 Racks/Cs.
    For Use With (Equipment) Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head
    Material Polypropylene
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.