missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Clear Tips, Nonsterile
Clear tips for accurate sample delivery
£154.00 - £252.00
Specifications
Filtered | Non-Filtered |
---|---|
Sterility | Non-sterile |
Color | Clear |
Autoclavable | Autoclavable |
Certifications/Compliance | Non-ANSI |
Product Code | Brand | Color | Sterility | Volume (Metric) | Format | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Sterility | Volume (Metric) | Format | Price | Quantity & Availability | ||
11344295
|
Axygen™
TT-200-C-HTR |
Clear | Non-sterile | 200 μL | Rack |
£231.00
Pack of 2304 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
11394295
|
Axygen™
TT-50-C-HTR |
Clear | Non-sterile | 50 μL | Rack |
£252.00
Pack of 2304 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
11344285
|
Axygen™
TT-1000-C-HTR |
Clear | Non-sterile | 1000 μL | Rack |
£154.00
Pack of 1536 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||
11394285
|
Axygen™
TT-20-C-HTR |
Clear | Non-sterile | 20 μL | Rack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Axygen™ offers a comprehensive line of tips for Tecan™, Aurora Biomed and PerkinElmer™ automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in nonsterile, sterile, non-filtered and filtered. Clear and Conductive black polypropylene tips are available in 96-well format and is also available in hanging tip rack format designed for Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head and PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head.Certifications
Certified DNAse-, RNAse- and Pyrogen-safe
Specifications
Non-Filtered | |
Clear | |
Non-ANSI | |
Rack | |
Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head |
Non-sterile | |
Autoclavable | |
For accurate sample delivery | |
Automation, Round | |
Polypropylene |