missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Clear Tips, Nonsterile
Clear tips for accurate sample delivery
Brand: Axygen TT-200-C-HTR
Code : 68
Additional Details : Weight : 4.10000kg
Description
- Polypropylene
- Volume ranging from 20 to 1000μL
- Available with or without aerosol-resistant filters
- Compatible with Axygen™ Tecan™/Perkin Elmer/Aurora™ Biomed/Corbett Cas-1200 systems
Specifications
Robotic Tip | |
Nonsterile | |
Yes | |
Round | |
Rack | |
Non-ANSI | |
Tecan™ Genesis™ Freedom™/Freedom Evo™/Miniprep with LiHa-1, 4, 8 head, Aurora Biomed Versa™ with 8-channel head |
200μL | |
Clear | |
Racked | |
Polypropylene | |
96 Tips/Rack | |
For accurate sample delivery |