Learn More
Agilent BioTek Half-Size Dichroic Mirror, Ex. 415 to 540, Em. 560 to 850
Product Code. 15533553 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Cytation 5, Synergy H1, Synergy LX models
Unit Size:
Each
Product Code. 15533553
Brand: Agilent BioTek 7138550
For use with Cytation 5, Synergy H1, Synergy LX models
  • Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cytation 5, Synergy H1, Synergy LX models