missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Half Fraser Broth

Product Code. 13265469
Click to view available options
Description:
Half Fraser Broth
Quantity:
10/Pk.
Fill Volume:
225 mL
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 13265469

Brand: Thermo Scientific™ BO0350S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate Listeria spp. from food and environmental samples with Thermo Scientific™ Half Fraser Broth enrichment medium.

Isolate Listeria spp. from food and environmental samples with Thermo Scientific™ Half Fraser Broth enrichment medium. Half Fraser Broth is a secondary enrichment broth based on the formula described by Fraser and Sperber1. Blackening of the medium is presumptive evidence of the presence of Listeria. All Half Fraser Broth enrichment cultures should be subcultured to plating medium.

Half Fraser Broth enrichment medium is intended for the isolation of Listeria spp. from food and environmental samples when used as the secondary enrichment medium according to ISO 11290-1 for Listeria isolation.

The medium contains ferric ammonium citrate and lithium chloride. All Listeria spp. hydrolyse aesculin to aesculetin. Aesculetin reacts with ferric ions which results in blackening. Another possible advantage to the addition of ferric ammonium citrate is that it has been shown that ferric ions enhance the growth of Listeria monocytogenes2. Lithium chloride is included in the medium to inhibit the growth of enterococci which can also hydrolyse aesculin. Blackening of the medium is presumptive evidence of the presence of Listeria. Contrary to early indications, cultures which do not blacken cannot be assumed to be Listeria-free. Therefore, all Half Fraser Broth enrichment cultures should be sub-cultured to plating medium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw with blue iridescence
Content And Storage 2°C to 12°C
Description Half Fraser Broth
Format Sirop Bottle
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Volume 225 mL
Fill Weight 225 ± 4mL
Packaging Type 10 bottles in a carton box
Quantity 10/Pk.

For laboratory use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.