Learn More
Scientific Glass Labs™ Simax Borosilicate Glass Evaporating Basin
Borosilicate glass construction with round bottom and spout.
Brand: Scientific Glass Labs™ 177/80
Code : 01
Additional Details : CAS Number : 175203-51-7 Weight : 1.00000kg
Description
- Borosilicate glass construction
- Clear
- Round bottom with spout
- Supplied in packs of 10
Specifications
Round | |
Borosilicate Glass | |
Evaporating Basin | |
39 mm |
100 mL | |
Round | |
Up to 400°C | |
85 mm |