missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAGH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | HAGH |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16827054
|
Proteintech
17196-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16817054
|
Proteintech
17196-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thiolesterase that catalyzes the hydrolysis of S-D-lactoyl-glutathione to form glutathione and D-lactic acid.Specifications
| HAGH | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| O35952, Q16775, Q99KB8 | |
| 14651, 24439, 3029 | |
| HAGH Fusion Protein Ag9998 | |
| Primary | |
| -20°C | |
| HAGH |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| GLO2, Glx II, GLX2, GLXII, Glyoxalase II, HAGH, HAGH1 | |
| Hagh | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title