missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAGH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17196-1-AP-150UL
This item is not returnable.
View return policy
Description
Thiolesterase that catalyzes the hydrolysis of S-D-lactoyl-glutathione to form glutathione and D-lactic acid.Specifications
| HAGH | |
| Polyclonal | |
| Unconjugated | |
| HAGH | |
| GLO2, Glx II, GLX2, GLXII, Glyoxalase II, HAGH, HAGH1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14651, 24439, 3029 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35952, Q16775, Q99KB8 | |
| Hagh | |
| HAGH Fusion Protein Ag9998 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction