missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Haemophilus Test Medium (HTM)

Product Code. 10536773
This item is not returnable. View return policy
Product Code. 10536773 Supplier Thermo Scientific™ Supplier No. PO5138A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform effortless susceptibility testing of Haemophilus influenza with this ready-to-use medium.

Perform effortless susceptibility testing of Haemophilus influenza with ready-to-use Thermo Scientific™ Oxoid™ Haemophilus Test Medium (HTM). The medium forms part of the recommended methods of the Clinical and Laboratory Standards Institute (CLSI)1. Developed from Mueller-Hinton Chocolate Agar, the medium has been found to be a highly reproducible medium for Haemophilus susceptibility testing2, 3. The transparency of the medium allows zones of inhibition to be easily read through the bottom of the plate.

Use Haemophilus Test Medium (HTM) for the susceptibility testing of Haemophilus influenzae and Haemophilus parainfluenzae.

  • Conforms to CLSI standards
  • Produces highly reproducible results.
  • Transparent medium to facilitate the reading of zones of inhibition.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Sand yellow
Content And Storage 2° to 12°C, in the dark
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar
Fill Weight 20g ± 5%
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.