Learn More
Thermo Scientific™ Haemophilus Selective Agar (Chocolate Agar with Vitox, Bacitracin, Vancomycin, Amphotericin B)
Selectively isolate Haemophilus species from specimens containing mixed flora using Thermo Scientific™ Haemophilus Selective Agar prepared media plates.
Brand: Thermo Scientific™ PO5097A
36.60 GBP valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Additional Details : Weight : 0.30000kg
Description
Selectively isolate Haemophilus species from clinical specimens containing mixed flora using Thermo Scientific™ Haemophilus Selective Agar (Chocolate Agar with Vitox, Bacitracin, Vancomycin, Amphotericin B).
Haemophilus influenza requires components of erythrocytes for growth, designated as X and V factors, a condition which led to the naming of the genus Haemophilus. The X factor corresponds to the haemin components of haemoglobin, and V factor refers to NAD (Nicotinamide-adenine-dinucleotide). In addition to a rich nutrient offering, Haemophilus Selective Agar contains both growth factors, as well as selective additions for inhibition of the accompanying flora from clinical materials. Bacitracin and Vancomycin suppress primarily gram-positive organisms, while Amphotericin B is effective against yeasts and fungi.Specifications
6°C to 12°C | |
90mm Monoplate | |
17.5g ± 5% | |
10/Pk. |
Haemophilus Selective Agar | |
Agar | |
Wrapped in single cellulose-based film wrap |